missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Octadecanol, 95%, Thermo Scientific Chemicals
$106.82 - $106.82
Chemical Identifiers
CAS | 112-92-5 |
---|---|
Molecular Formula | C18H38O |
Molecular Weight (g/mol) | 270.5 |
MDL Number | MFCD00002823 |
InChI Key | GLDOVTGHNKAZLK-UHFFFAOYSA-N |
Synonym | stearyl alcohol, 1-octadecanol, octadecanol, octadecyl alcohol, n-octadecanol, 1-hydroxyoctadecane, n-1-octadecanol, stearol, n-octadecyl alcohol, stearic alcohol |
PubChem CID | 8221 |
ChEBI | CHEBI:32154 |
IUPAC Name | octadecan-1-ol |
SMILES | CCCCCCCCCCCCCCCCCCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC129300010
|
Thermo Scientific Chemicals
129300010 |
1 kg | Plastic bottle |
Each for $106.82
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-92-5 | |
270.5 | |
GLDOVTGHNKAZLK-UHFFFAOYSA-N | |
8221 | |
octadecan-1-ol |
C18H38O | |
MFCD00002823 | |
stearyl alcohol, 1-octadecanol, octadecanol, octadecyl alcohol, n-octadecanol, 1-hydroxyoctadecane, n-1-octadecanol, stearol, n-octadecyl alcohol, stearic alcohol | |
CHEBI:32154 | |
CCCCCCCCCCCCCCCCCCO |
Specifications
112-92-5 | |
94.0 | |
55.0°C to 58.0°C | |
185g/mL | |
185°C | |
94% min. (GC) | |
C18H38O | |
MFCD00002823 | |
01, 431 | |
stearyl alcohol, 1-octadecanol, octadecanol, octadecyl alcohol, n-octadecanol, 1-hydroxyoctadecane, n-1-octadecanol, stearol, n-octadecyl alcohol, stearic alcohol | |
GLDOVTGHNKAZLK-UHFFFAOYSA-N | |
octadecan-1-ol | |
8221 | |
270.5 | |
Flakes |
1mg KOH/g max. | |
100.0 | |
White | |
210.0°C (15.0 mmHg) | |
Authentic | |
Plastic bottle | |
CH3(CH2)17OH | |
1 kg | |
15, 8932 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and benzene | |
CCCCCCCCCCCCCCCCCCO | |
270.5 | |
CHEBI:32154 | |
95% | |
1-Octadecanol |
Safety and Handling
EINECSNumber : 204-017-6
RUO – Research Use Only