missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Octadecene, 90%, tech., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 129310025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Octadecene | |
17.5°C | |
0.7890g/mL | |
154°C | |
90% | |
C18H36 | |
CH3(CH2)15CH=CH2 | |
01,226 | |
1-octadecene, octadecene, alpha-octadecene, .alpha.-octadecene, unii-h5zuq6v4ak, octadecene-1, h5zuq6v4ak, alkenes, c14-20 .alpha., 1-octadecene, analytical standard, linear octadecene | |
CCCMONHAUSKTEQ-UHFFFAOYSA-N | |
octadec-1-ene | |
8217 | |
CHEBI:30824 | |
90% |
112-88-9 | |
Colorless to Amber | |
315°C | |
Authentic | |
Glass bottle | |
1.4435 to 1.4455 | |
2.5 L | |
0.789 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether, petroleum and, coal-tar solvents | |
CCCCCCCCCCCCCCCCC=C | |
252.48 | |
3.6 mm2/s (40°C) | |
252.48 | |
Liquid |
Chemical Identifiers
112-88-9 | |
252.48 | |
1-octadecene, octadecene, alpha-octadecene, .alpha.-octadecene, unii-h5zuq6v4ak, octadecene-1, h5zuq6v4ak, alkenes, c14-20 .alpha., 1-octadecene, analytical standard, linear octadecene | |
CHEBI:30824 | |
CCCCCCCCCCCCCCCCC=C |
C18H36 | |
CCCMONHAUSKTEQ-UHFFFAOYSA-N | |
8217 | |
octadec-1-ene |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
GHS Signal Word: Danger
EINECSNumber : 204-012-9
RUO – Research Use Only