Learn More
1-Octadecene, tech. 90%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L11004AU
Description
1-Octadecene is an inexpensive solvent having high boiling point. Due to this property, it replaces the heat transfer fluids such as dowthem A or therminol 66 and used in the synthesis of high quality CdSe quantum dots (nano crystal made up of semiconductor materials). These quantum dots find application in transistors, diode lasers and solar cells. It is also used as a solvent for the synthesis of iron oxide nanospheres.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Octadecene is an inexpensive solvent having high boiling point. Due to this property, it replaces the heat transfer fluids such as dowthem A or therminol 66 and used in the synthesis of high quality CdSe quantum dots (nano crystal made up of semiconductor materials). These quantum dots find application in transistors, diode lasers and solar cells. It is also used as a solvent for the synthesis of iron oxide nanospheres.
Solubility
Miscible with alcohol, acetone and ether. Immiscible with water.
Notes
Avoid reaction with strong Lewis or mineral acids. Free radical initiators should be avoided. Keep container tightly closed in a dry and well-ventilated place.
Specifications
1-Octadecene | |
15°C to 17°C | |
178°C to 179°C (15 mmHg) | |
C18H36 | |
MFCD00009003 | |
1761067 | |
Miscible with alcohol,acetone and ether. Immiscible with water. | |
CCCCCCCCCCCCCCCCC=C | |
252.486 | |
CHEBI:30824 | |
90% |
112-88-9 | |
0.789 | |
154°C (309°F) | |
1.444 | |
1000 mL | |
1-octadecene, octadecene, alpha-octadecene, .alpha.-octadecene, unii-h5zuq6v4ak, octadecene-1, h5zuq6v4ak, alkenes, c14-20 .alpha., 1-octadecene, analytical standard, linear octadecene | |
CCCMONHAUSKTEQ-UHFFFAOYSA-N | |
octadec-1-ene | |
8217 | |
252.48 | |
Technical |
Chemical Identifiers
112-88-9 | |
252.486 | |
CCCMONHAUSKTEQ-UHFFFAOYSA-N | |
8217 | |
octadec-1-ene |
C18H36 | |
MFCD00009003 | |
1-octadecene, octadecene, alpha-octadecene, .alpha.-octadecene, unii-h5zuq6v4ak, octadecene-1, h5zuq6v4ak, alkenes, c14-20 .alpha., 1-octadecene, analytical standard, linear octadecene | |
CHEBI:30824 | |
CCCCCCCCCCCCCCCCC=C |
Safety and Handling
P301+P310-P331-P501c
H304
EINECSNumber : 204-012-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only