Learn More
1-Octadecylamine hydrochloride, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04137814
Description
1-Octadecylamine hydrochloride is used as organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Octadecylamine hydrochloride | |
160°C | |
MFCD00042018 | |
octadecylamine hydrochloride, stearylamine hydrochloride, 1-octadecylamine hydrochloride, stearamine hydrochloride, unii-3v3v50q0sj, 1-octadecanamine, hydrochloride, octadecylamine, hydrochloride, acmc-1bnqe, octadecyl-ammonium chloride, c18h39n.hcl | |
RNYJXPUAFDFIQJ-UHFFFAOYSA-N | |
octadecan-1-amine;hydrochloride | |
519552 | |
Powder |
1838-08-0 | |
C18H40ClN | |
25 g | |
Soluble in hot methanol. | |
CCCCCCCCCCCCCCCCCCN.Cl | |
305.975 | |
305.97 |
Chemical Identifiers
1838-08-0 | |
305.975 | |
RNYJXPUAFDFIQJ-UHFFFAOYSA-N | |
519552 | |
CCCCCCCCCCCCCCCCCCN.Cl |
C18H40ClN | |
MFCD00042018 | |
octadecylamine hydrochloride, stearylamine hydrochloride, 1-octadecylamine hydrochloride, stearamine hydrochloride, unii-3v3v50q0sj, 1-octadecanamine, hydrochloride, octadecylamine, hydrochloride, acmc-1bnqe, octadecyl-ammonium chloride, c18h39n.hcl | |
octadecan-1-amine;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 217-410-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only