missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1 Octane Sulfonic Acid Sodium Salt, HPLC Ion Pair Agent, Fisher Chemical™

Improve HPLC separation of basic analytes by ion pairing.

Supplier:  Thermo Fisher Scientific O002825

 View more versions of this product

Catalog No. O002825


Explore available promotions

Description

Description

  • High purity
  • Application tested
  • Dissolves easily
Specifications

Specifications

5324-84-5
1.5% Max.
C8H17NaO3S
25 g
[Na+].CCCCCCCCS([O-])(=O)=O
216.27
HPLC
≤ 1.00 AU at 210 nm, ≤ 0.1 AU at 220 nm, ≤ 0.04 AU at 254 nm, ≤ 0.04 AU at 280nm
Conforms
MFCD00007544
HRQDCDQDOPSGBR-UHFFFAOYSA-M
sodium octane-1-sulfonate
216.7
Powder

Chemical Identifiers

5324-84-5
216.27
HRQDCDQDOPSGBR-UHFFFAOYSA-M
[Na+].CCCCCCCCS([O-])(=O)=O
C8H17NaO3S
MFCD00007544
sodium octane-1-sulfonate
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient

SDS Documents

Documents

Promotions

Promotions