Learn More
1-Octanesulfonic Acid, Sodium Salt Anhydrous, HPLC Grade, Thermo Scientific Chemicals
$1396.14 - $1396.14
Chemical Identifiers
CAS | 5324-84-5 |
---|---|
Molecular Formula | C8H17NaO3S |
Molecular Weight (g/mol) | 216.27 |
MDL Number | MFCD00007544 |
InChI Key | HRQDCDQDOPSGBR-UHFFFAOYSA-M |
Synonym | sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8 |
PubChem CID | 23669624 |
IUPAC Name | sodium octane-1-sulfonate |
SMILES | [Na+].CCCCCCCCS([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC416361000
|
Thermo Scientific Chemicals
416361000 |
100 g |
Each for $1,396.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatographyChemical Identifiers
5324-84-5 | |
216.27 | |
HRQDCDQDOPSGBR-UHFFFAOYSA-M | |
23669624 | |
[Na+].CCCCCCCCS([O-])(=O)=O |
C8H17NaO3S | |
MFCD00007544 | |
sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8 | |
sodium octane-1-sulfonate |
Specifications
5324-84-5 | |
>300°C | |
1.5% max. (1 g, 110°C, 75 min) | |
C8H17NaO3S | |
MFCD00007544 | |
04, III, 25 | |
Solubility in water: freely soluble. Other solubilities: 4g/L in methanol | |
[Na+].CCCCCCCCS([O-])(=O)=O | |
216.27 | |
216.28 | |
Crystalline Solid |
(2.7 g/50 ml water) (Acidified with ACS grade Phosphoric Acid to pH 3), 0.04 max. at 254nm, 0.04 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm | |
White | |
Authentic | |
CH3(CH2)7SO3Na | |
100 g | |
sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8 | |
HRQDCDQDOPSGBR-UHFFFAOYSA-M | |
sodium octane-1-sulfonate | |
23669624 | |
HPLC | |
1-Octanesulfonic Acid, Sodium Salt |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 226-195-4
TSCA : TSCA
RUO – Research Use Only