missing translation for 'onlineSavingsMsg'
Learn More

1-Octen-3-ol, 98%

CAS: 3391-86-4 | C8H16O | 128.22 g/mol

$44.49 - $343.14

Chemical Identifiers

CAS 3391-86-4
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00004589
InChI Key VSMOENVRRABVKN-UHFFFAOYSA-N
Synonym 1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol
PubChem CID 18827
ChEBI CHEBI:34118
IUPAC Name oct-1-en-3-ol
SMILES CCCCCC(C=C)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC129470050
SDS View Documents
Thermo Scientific Chemicals
129470050
5 g Glass bottle
Each for $44.49
Only null left
 
AC129470250
SDS View Documents
Thermo Scientific Chemicals
129470250
25 g Glass bottle
Each for $120.66
Only null left
 
AC129471000
SDS View Documents
Thermo Scientific Chemicals
129471000
100 g Glass bottle
Each for $343.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3391-86-4
128.22
VSMOENVRRABVKN-UHFFFAOYSA-N
18827
oct-1-en-3-ol
C8H16O
MFCD00004589
1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol
CHEBI:34118
CCCCCC(C=C)O

Specifications

3391-86-4
100.0
0.8370g/mL
68°C
97.5% min. (GC)
C8H16O
CH3(CH2)4CH(OH)CH=CH2
5 g
1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol
VSMOENVRRABVKN-UHFFFAOYSA-N
oct-1-en-3-ol
18827
128.22
Liquid
97.5
Colorless to Yellow
174.0°C
Authentic
Glass bottle
1.4340 to 1.4420
MFCD00004589
0.837
Solubility in water: insoluble in water. Other solubilities: soluble in ethanol
CCCCCC(C=C)O
128.22
CHEBI:34118
98%
1-Octen-3-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 222-226-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only