missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Octen-3-one 95.0+%, TCI America™

Supplier:  TCI America O03821G

 View more versions of this product

Catalog No. O03821G


Explore available promotions

Description

Description

Specifications

Specifications

1-Octen-3-one
Yellow
C8H14O
1 g
1-octen-3-one, vinyl amyl ketone, amyl vinyl ketone, pentyl vinyl ketone, unii-7lt7z4q9xr, 1-octene-3-one, fema no. 3515, 7lt7z4q9xr, n-pentyl vinyl ketone, octen-3-one, 1
CCCCCC(=O)C=C
126.199
126.20
Liquid
4312-99-6
60°C
MFCD00036558
1224
KLTVSWGXIAYTHO-UHFFFAOYSA-N
oct-1-en-3-one
61346
≥95.0% (GC)

Chemical Identifiers

4312-99-6
126.199
KLTVSWGXIAYTHO-UHFFFAOYSA-N
61346
CCCCCC(=O)C=C
C8H14O
MFCD00036558
1-octen-3-one, vinyl amyl ketone, amyl vinyl ketone, pentyl vinyl ketone, unii-7lt7z4q9xr, 1-octene-3-one, fema no. 3515, 7lt7z4q9xr, n-pentyl vinyl ketone, octen-3-one, 1
oct-1-en-3-one
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions