missing translation for 'onlineSavingsMsg'
Learn More

1-Octene, 99+%

CAS: 111-66-0 | C8H16 | 112.21 g/mol

$97.69 - $693.87

Chemical Identifiers

CAS 111-66-0
Molecular Formula C8H16
Molecular Weight (g/mol) 112.21
MDL Number MFCD00009548
InChI Key KWKAKUADMBZCLK-UHFFFAOYSA-N
Synonym 1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene
PubChem CID 8125
ChEBI CHEBI:46708
IUPAC Name oct-1-ene
SMILES CCCCCCC=C
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC301251000
SDS View Documents
Thermo Scientific Chemicals
301251000
100 mL Glass bottle
Each for $97.69
Only null left
 
AC301255000
SDS View Documents
Thermo Scientific Chemicals
301255000
500 mL Glass bottle
Each for $112.42
Only null left
 
AC301250010
SDS View Documents
Thermo Scientific Chemicals
301250010
1 L Glass bottle
Each for $159.75
Only null left
 
AC301250025
SDS View Documents
Thermo Scientific Chemicals
301250025
2.5 L Glass bottle
Each for $353.80
Only null left
 
AC301250100
SDS View Documents
Thermo Scientific Chemicals
301250100
10 L Metal Drum
Each for $693.87
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111-66-0
112.21
KWKAKUADMBZCLK-UHFFFAOYSA-N
8125
oct-1-ene
C8H16
MFCD00009548
1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene
CHEBI:46708
CCCCCCC=C

Specifications

111-66-0
100.0
0.7150g/mL
13°C
99% min. (GC)
C8H16
CH3(CH2)5CH=CH2
100 mL
0.715
1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene
KWKAKUADMBZCLK-UHFFFAOYSA-N
oct-1-ene
8125
CHEBI:46708
99+%
1-Octene, 99+%
99.0
-102.0°C
121.0°C to 123.0°C
Authentic
Glass bottle
1.4080 to 1.4100
MFCD00009548
01, 221
15, 1766
Solubility in water: 4.1 mg/l (25°C). Other solubilities: soluble in alcohol, acetone, ether, petroleum and, coal-tar solvents, soluble in hydrocarbon solvents
CCCCCCC=C
112.21
0.38 mm2/s (40°C)
112.21
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.
Very toxic to aquatic life with long lasting effects.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environm

GHS Signal Word: Danger

EINECSNumber : 203-893-7

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only