missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Octylphosphonic dichloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1858714

 View more versions of this product

Catalog No. AAL1858714


Explore available promotions

Description

Description

1-Octylphosphonic dichloride is employed in the surface modification of calcium fluoro and hydroxyapatite by 1-octylphosphonic dichloride and qualitative and quantitative analysis of phosphate rock from Hazm Al-jalamid area, northern Saudi Arabia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octylphosphonic dichloride is employed in the surface modification of calcium fluoro and hydroxyapatite by 1-octylphosphonic dichloride and qualitative and quantitative analysis of phosphate rock from Hazm Al-jalamid area, northern Saudi Arabia.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from strong bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Specifications

1-Octylphosphonic dichloride
1.115
C8H17Cl2OP
MFCD00015813
UN3265
Moisture sensitive
Reacts with water.
CCCCCCCCP(=O)(Cl)Cl
231.097
231.1
3095-94-1
143°C to 145°C (12 mmHg)
1.4665
25 g
1931202
octylphosphonic dichloride, 1-octylphosphonic dichloride, octylphosphonoyl dichloride, acmc-1afrz, octyl phosphonic dichloride, dichloro octyl phosphine oxide, phosphonic dichloride,p-octyl
RTHCRCDXILKTCY-UHFFFAOYSA-N
1-dichlorophosphoryloctane
76541
97%

Chemical Identifiers

3095-94-1
231.097
RTHCRCDXILKTCY-UHFFFAOYSA-N
76541
CCCCCCCCP(=O)(Cl)Cl
C8H17Cl2OP
MFCD00015813
octylphosphonic dichloride, 1-octylphosphonic dichloride, octylphosphonoyl dichloride, acmc-1afrz, octyl phosphonic dichloride, dichloro octyl phosphine oxide, phosphonic dichloride,p-octyl
1-dichlorophosphoryloctane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 221-444-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only