Learn More
1-Octyne, 98%, Thermo Scientific Chemicals
$64.29 - $332.37
Chemical Identifiers
CAS | 629-05-0 |
---|---|
Molecular Formula | C8H14 |
Molecular Weight (g/mol) | 110.20 |
MDL Number | MFCD00009546 |
InChI Key | UMIPWJGWASORKV-UHFFFAOYSA-N |
Synonym | 1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g |
PubChem CID | 12370 |
IUPAC Name | oct-1-yne |
SMILES | CCCCCCC#C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1244009
|
Thermo Scientific Chemicals
A1244009 |
10 g |
N/A
|
|
||||||
AAA1244014
|
Thermo Scientific Chemicals
A1244014 |
25 g |
N/A
|
|
||||||
AAA1244022
|
Thermo Scientific Chemicals
A1244022 |
100 g |
N/A
|
|
||||||
Description
1-Octyne is widely used in organic reactions such as halogenation, hydration, oxidative cleavage, hydrogenation, hydrohalogenation, nitrile formation, polymerization and substitution reactions. It is used a precursor for the preparation of monohalogen substituted akenes or dihalogen substituted alkanes on treated with hydrogen halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Octyne is widely used in organic reactions such as halogenation, hydration, oxidative cleavage, hydrogenation, hydrohalogenation, nitrile formation, polymerization and substitution reactions. It is used a precursor for the preparation of monohalogen substituted akenes or dihalogen substituted alkanes on treated with hydrogen halides.
Solubility
Miscible with alcohol, ether and other organic solvents. Immiscible with water.
Notes
Incompatible with oxidizing agents. Keep away from heat, sparks, sources of ignition and flame.
Chemical Identifiers
629-05-0 | |
110.20 | |
UMIPWJGWASORKV-UHFFFAOYSA-N | |
12370 | |
CCCCCCC#C |
C8H14 | |
MFCD00009546 | |
1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g | |
oct-1-yne |
Specifications
629-05-0 | |
0.746 | |
17°C (62°F) | |
C8H14 | |
CH3(CH2)5C≡CH | |
10 g | |
1734494 | |
Miscible with alcohol,ether and other organic solvents. Immiscible with water. | |
CCCCCCC#C | |
110.20 | |
110.2 | |
Liquid |
-60°C | |
123°C to 126°C | |
98% | |
1.416 | |
MFCD00009546 | |
UN3295 | |
1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g | |
UMIPWJGWASORKV-UHFFFAOYSA-N | |
oct-1-yne | |
12370 | |
98% | |
1-Octyne |
Safety and Handling
GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
H225-H304
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 211-069-3
RTECSNumber : RI2655000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only