missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(p-Toluenesulfonyl)indole, 95%
CAS: 31271-90-6 | C15H13NO2S | 271.33 g/mol
$127.27 - $1,182.75
Chemical Identifiers
| CAS | 31271-90-6 |
|---|---|
| Molecular Formula | C15H13NO2S |
| Molecular Weight (g/mol) | 271.33 |
| MDL Number | MFCD01165032 |
| InChI Key | JNRRPYFLDADLJW-UHFFFAOYSA-N |
| Synonym | 1-tosyl-1h-indole, n-tosylindole, 1-p-toluenesulfonyl indole, 1-4-methylbenzenesulfonyl indole, n-p-toluenesulfonyl indole, 1-4-methylphenyl sulfonyl-1h-indole, 1-tosylindole, 1-p-tolylsulfonyl indole, 1-p-toluenesulphonyl indole, 4-methylphenyl sulfonyl indole |
| PubChem CID | 10956616 |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)N1C=CC2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2232003
|
Thermo Scientific Chemicals
B2232003 |
1 g |
N/A
|
|
|||||
|
AAB2232006
|
Thermo Scientific Chemicals
B2232006 |
5 g |
N/A
|
|
|||||
|
AAB2232014
|
Thermo Scientific Chemicals
B2232014 |
25 g |
N/A
|
|
|||||
Description
1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 31271-90-6 | |
| 271.33 | |
| JNRRPYFLDADLJW-UHFFFAOYSA-N | |
| 10956616 |
| C15H13NO2S | |
| MFCD01165032 | |
| 1-tosyl-1h-indole, n-tosylindole, 1-p-toluenesulfonyl indole, 1-4-methylbenzenesulfonyl indole, n-p-toluenesulfonyl indole, 1-4-methylphenyl sulfonyl-1h-indole, 1-tosylindole, 1-p-tolylsulfonyl indole, 1-p-toluenesulphonyl indole, 4-methylphenyl sulfonyl indole | |
| CC1=CC=C(C=C1)S(=O)(=O)N1C=CC2=CC=CC=C12 |
Specifications
| 31271-90-6 | |
| C15H13NO2S | |
| 1 g | |
| Insoluble in water. | |
| CC1=CC=C(C=C1)S(=O)(=O)N1C=CC2=CC=CC=C12 | |
| 271.33 | |
| 271.33 | |
| 1-(p-Toluenesulfonyl)indole |
| 84°C to 86°C | |
| MFCD01165032 | |
| 1-tosyl-1h-indole, n-tosylindole, 1-p-toluenesulfonyl indole, 1-4-methylbenzenesulfonyl indole, n-p-toluenesulfonyl indole, 1-4-methylphenyl sulfonyl-1h-indole, 1-tosylindole, 1-p-tolylsulfonyl indole, 1-p-toluenesulphonyl indole, 4-methylphenyl sulfonyl indole | |
| JNRRPYFLDADLJW-UHFFFAOYSA-N | |
| 1-(4-methylphenyl)sulfonylindole | |
| 10956616 | |
| 95% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only