missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
1-Pentanol | |
-79°C | |
137°C | |
MFCD00002977,MFCD00081734,MFCD01075169 | |
1105 | |
AMQJEAYHLZJPGS-UHFFFAOYSA-N | |
pentan-1-ol | |
6276 | |
88.15 | |
Liquid |
71-41-0 | |
Colorless | |
C5H12O | |
500 mL | |
1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol | |
CCCCCO | |
88.15 | |
CHEBI:44884 | |
≥99.0% (GC) |
Chemical Identifiers
71-41-0 | |
88.15 | |
AMQJEAYHLZJPGS-UHFFFAOYSA-N | |
6276 | |
pentan-1-ol |
C5H12O | |
MFCD00002977,MFCD00081734,MFCD01075169 | |
1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol | |
CHEBI:44884 | |
CCCCCO |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : SB9800000
TSCA : Yes