missing translation for 'onlineSavingsMsg'
Learn More

1-Penten-3-ol, 97%

CAS: 616-25-1 | C5H10O | 86.13 g/mol

$68.14 - $190.66

Chemical Identifiers

CAS 616-25-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00004573
InChI Key VHVMXWZXFBOANQ-UHFFFAOYNA-N
Synonym 1-penten-3-ol, ethyl vinyl carbinol, vinyl ethyl carbinol, 1-pentene-3-ol, 1-ethylallyl alcohol, alpha-ethylallyl alcohol, 1-penten-3-ol e, 1-penten-3-ol, r, fema no. 3584, ethylvinylcarbinol
PubChem CID 12020
IUPAC Name pent-1-en-3-ol
SMILES CCC(O)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAB2015506
View Documents
Thermo Scientific Chemicals
B2015506
5 g
Each for $68.14
Only null left
 
AAB2015514
View Documents
Thermo Scientific Chemicals
B2015514
25 g
Each for $190.66
Only null left
 
Description

Description

1-Penten-3-ol is used as a flavor and a fragrance. It can be used for lift with cucumber, melon, berry, horseradish, strawberry, and tomato.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Penten-3-ol is used as a flavor and a fragrance. It can be used for lift with cucumber, melon, berry, horseradish, strawberry, and tomato.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications

Chemical Identifiers

616-25-1
86.13
VHVMXWZXFBOANQ-UHFFFAOYNA-N
12020
CCC(O)C=C
C5H10O
MFCD00004573
1-penten-3-ol, ethyl vinyl carbinol, vinyl ethyl carbinol, 1-pentene-3-ol, 1-ethylallyl alcohol, alpha-ethylallyl alcohol, 1-penten-3-ol e, 1-penten-3-ol, r, fema no. 3584, ethylvinylcarbinol
pent-1-en-3-ol

Specifications

616-25-1
114°C to 115°C
C5H10O
MFCD00004573
UN1987
1-penten-3-ol, ethyl vinyl carbinol, vinyl ethyl carbinol, 1-pentene-3-ol, 1-ethylallyl alcohol, alpha-ethylallyl alcohol, 1-penten-3-ol e, 1-penten-3-ol, r, fema no. 3584, ethylvinylcarbinol
VHVMXWZXFBOANQ-UHFFFAOYNA-N
pent-1-en-3-ol
12020
≥98%
0.838
25°C (77°F)
1.425
5 g
1719834
Slightly soluble in water.
CCC(O)C=C
86.13
86.13
1-Penten-3-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 210-472-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only