Learn More
1-Pentene, 97%, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 430731000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Pentene | |
96.0 | |
-165.0°C | |
0.6400g/mL | |
−28°C | |
96% min. (GC) | |
C5H10 | |
CH3CH2CH2CH=CH2 | |
100 mL | |
15,7234 | |
Solubility in water: 0.15g/L (20°C). Other solubilities: miscible with alcohol, ether and beznene, soluble in most organic solvents | |
CCCC=C | |
70.14 | |
0.202 mPa.s (20°C) | |
97% |
109-67-1 | |
100.0 | |
Colorless | |
30.0°C | |
Authentic | |
AcroSeal™ Glass Bottle | |
1.3704 to 1.3724 | |
MFCD00003567 | |
0.64 | |
1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1 | |
YWAKXRMUMFPDSH-UHFFFAOYSA-N | |
pent-1-ene | |
8004 | |
70.13 | |
Liquid |
Chemical Identifiers
109-67-1 | |
70.14 | |
YWAKXRMUMFPDSH-UHFFFAOYSA-N | |
8004 | |
CCCC=C |
C5H10 | |
MFCD00003567 | |
1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1 | |
pent-1-ene |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
Extremely flammable liquid and vapor.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Ta
GHS Signal Word: Danger
EINECSNumber : 203-694-5
RUO – Research Use Only