missing translation for 'onlineSavingsMsg'
Learn More

1-Pentene, 97%

CAS: 109-67-1 | C5H10 | 70.14 g/mol

$119.80 - $219.52

Chemical Identifiers

CAS 109-67-1
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00003567
InChI Key YWAKXRMUMFPDSH-UHFFFAOYSA-N
Synonym 1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
PubChem CID 8004
IUPAC Name pent-1-ene
SMILES CCCC=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC268660250
SDS View Documents
Thermo Scientific Chemicals
268660250
25 mL Glass bottle
Each for $119.80
Only null left
 
AC268661000
SDS View Documents
Thermo Scientific Chemicals
268661000
100 mL Glass bottle
Each for $219.52
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

109-67-1
70.14
YWAKXRMUMFPDSH-UHFFFAOYSA-N
8004
CCCC=C
C5H10
MFCD00003567
1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
pent-1-ene

Specifications

109-67-1
100.0
Colorless
30.0°C
Authentic
Glass bottle
1.3704 to 1.3724
MFCD00003567
01,21
15,7234
Solubility in water: 0.15g/L (20°C). Other solubilities: miscible with alcohol, ether and beznene, soluble in most organic solvents
CCCC=C
70.14
0.202 mPa.s (20°C)
97%
1-Pentene
96.0
-165.0°C
0.6400g/mL
−28°C
97%
C5H10
CH3CH2CH2CH=CH2
25 mL
0.64
1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
YWAKXRMUMFPDSH-UHFFFAOYSA-N
pent-1-ene
8004
70.13
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be fatal if swallowed and enters airways.
Extremely flammable liquid and vapor.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Ta

GHS Signal Word: Danger

EINECSNumber : 203-694-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only