missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Pentene, 97%, Thermo Scientific Chemicals

$116.26 - $305.78

Chemical Identifiers

CAS 109-67-1
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00003567
InChI Key YWAKXRMUMFPDSH-UHFFFAOYSA-N
Synonym 1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
PubChem CID 8004
IUPAC Name pent-1-ene
SMILES CCCC=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1672314
View Documents
Thermo Scientific Chemicals
A1672314
25 g
Each for $116.26
 
AAA1672322
View Documents
Thermo Scientific Chemicals
A1672322
100 g
Each for $305.78
 
Description

Description

1-Pentene is used in the preparation of d-xylitol, l-arabinitol and hydrazones of substituted salicylaldehydes. It is also used as a blending agent for of high-octane gasoline. It reacts with p-toluenesulfinate to yield 1-(2-iodo-pentane-1-sulfonyl)-4-methyl-benzene. It is also involved in Gas Chromatography and Liquid Chromatography analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentene is used in the preparation of d-xylitol, l-arabinitol and hydrazones of substituted salicylaldehydes. It is also used as a blending agent for of high-octane gasoline. It reacts with p-toluenesulfinate to yield 1-(2-iodo-pentane-1-sulfonyl)-4-methyl-benzene. It is also involved in Gas Chromatography and Liquid Chromatography analysis.

Solubility
Miscible with ethanol, ethyl ether, benzene and organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, reducing agents, halogens and acids.
Specifications

Chemical Identifiers

109-67-1
70.14
YWAKXRMUMFPDSH-UHFFFAOYSA-N
8004
CCCC=C
C5H10
MFCD00003567
1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
pent-1-ene

Specifications

109-67-1
0.64
−51°C (−60°F)
1.3715
25 g
1731629
1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
YWAKXRMUMFPDSH-UHFFFAOYSA-N
pent-1-ene
8004
97%
-138°C
30°C to 31°C
C5H10
MFCD00003567
UN1108
14,7122
Miscible with ethanol,ethyl ether,benzene and organic solvents. Immiscible with water.
CCCC=C
70.14
70.14
1-Pentene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H224-H304
Extremely flammable liquid and vapour.
May be fatal if swallowed and enters airways.

P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c

H224-H304

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: 1-PENTENE

EINECSNumber : 203-694-5

RTECSNumber : SB2179000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only