Learn More
(1-Pentyl)triphenylphosphonium bromide, 98%
CAS: 21406-61-1 | C23H26BrP | 413.34 g/mol
$83.68 - $808.51
Chemical Identifiers
| CAS | 21406-61-1 |
|---|---|
| Molecular Formula | C23H26BrP |
| Molecular Weight (g/mol) | 413.34 |
| MDL Number | MFCD00031630 |
| InChI Key | VAUKWMSXUKODHR-UHFFFAOYSA-M |
| Synonym | pentyltriphenylphosphonium bromide, amyltriphenylphosphonium bromide, n-amyl triphenylphosphonium bromide, pentyltriphenylphosphanium bromide, 1-pentyl triphenylphosphonium bromide, triphenyl-n-amylphosphonium bromide, pentyl triphenyl phosphanium bromide, phosphonium, pentyltriphenyl-, bromide, pentyl triphenyl phosphonium bromide, pubchem15722 |
| PubChem CID | 2724565 |
| IUPAC Name | pentyl(triphenyl)phosphanium;bromide |
| SMILES | [Br-].CCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1253414
|
Thermo Scientific Chemicals
A1253414 |
25 g |
Each for $83.68
|
|
|||||
|
AAA1253422
|
Thermo Scientific Chemicals
A1253422 |
100 g |
Each for $229.49
|
|
|||||
|
AAA1253436
|
Thermo Scientific Chemicals
A1253436 |
500 g |
Each for $808.51
|
|
|||||
Description
It is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediates.
Solubility
Partly miscible in water. Soluble in methanol and chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
| 21406-61-1 | |
| 413.34 | |
| VAUKWMSXUKODHR-UHFFFAOYSA-M | |
| 2724565 | |
| [Br-].CCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C23H26BrP | |
| MFCD00031630 | |
| pentyltriphenylphosphonium bromide, amyltriphenylphosphonium bromide, n-amyl triphenylphosphonium bromide, pentyltriphenylphosphanium bromide, 1-pentyl triphenylphosphonium bromide, triphenyl-n-amylphosphonium bromide, pentyl triphenyl phosphanium bromide, phosphonium, pentyltriphenyl-, bromide, pentyl triphenyl phosphonium bromide, pubchem15722 | |
| pentyl(triphenyl)phosphanium;bromide |
Specifications
| 21406-61-1 | |
| C23H26BrP | |
| 25 g | |
| Hygroscopic | |
| Partly miscible in water. Soluble in methanol and chloroform. | |
| [Br-].CCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 413.34 | |
| 413.33 | |
| (1-Pentyl)triphenylphosphonium bromide |
| 164°C to 166°C | |
| MFCD00031630 | |
| 3580453 | |
| pentyltriphenylphosphonium bromide, amyltriphenylphosphonium bromide, n-amyl triphenylphosphonium bromide, pentyltriphenylphosphanium bromide, 1-pentyl triphenylphosphonium bromide, triphenyl-n-amylphosphonium bromide, pentyl triphenyl phosphanium bromide, phosphonium, pentyltriphenyl-, bromide, pentyl triphenyl phosphonium bromide, pubchem15722 | |
| VAUKWMSXUKODHR-UHFFFAOYSA-M | |
| pentyl(triphenyl)phosphanium;bromide | |
| 2724565 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 244-374-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only