missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Pentyne, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 303820050

 View more versions of this product

Catalog No. AC303820050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Pentyne
627-19-0
100.0
Colorless to Yellow
40.0°C
Authentic
Glass bottle
1.3840 to 1.3880
MFCD00009469
0.69
IBXNCJKFFQIKKY-UHFFFAOYSA-N
pent-1-yne
12309
99%
99%
98.5
-106°C to 105°C
0.6900g/mL
<−34°C
98.5% min. (GC)
C5H8
CH3CH2CH2C≡CH
5 g
1-pentyne, propylacetylene, pentyne, acetylene, propyl, pentin, pentyn, 1-n-pentyne, pent-1-yn-1-yl, acmc-1cuk3
CCCC#C
68.12
68.12
Liquid

Chemical Identifiers

627-19-0
68.12
IBXNCJKFFQIKKY-UHFFFAOYSA-N
12309
CCCC#C
C5H8
MFCD00009469
1-pentyne, propylacetylene, pentyne, acetylene, propyl, pentin, pentyn, 1-n-pentyne, pent-1-yn-1-yl, acmc-1cuk3
pent-1-yne
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-987-1

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only