missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1,2-propanedione, 98%

CAS: 579-07-7 | C9H8O2 | 148.16 g/mol

$65.06 - $65.06

Chemical Identifiers

CAS 579-07-7
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00008755
InChI Key BVQVLAIMHVDZEL-UHFFFAOYSA-N
Synonym 1-phenyl-1,2-propanedione, acetylbenzoyl, acetyl benzoyl, benzoylacetyl, methylphenylglyoxal, pyruvophenone, 1,2-propanedione, 1-phenyl, phenylmethyldiketone, benzoyl methyl ketone, methyl phenyl diketone
PubChem CID 11363
ChEBI CHEBI:63552
IUPAC Name 1-phenylpropane-1,2-dione
SMILES CC(=O)C(=O)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC207360010
SDS View Documents
Thermo Scientific Chemicals
207360010
1 g Glass bottle
Each for $65.06
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

579-07-7
148.16
BVQVLAIMHVDZEL-UHFFFAOYSA-N
11363
1-phenylpropane-1,2-dione
C9H8O2
MFCD00008755
1-phenyl-1,2-propanedione, acetylbenzoyl, acetyl benzoyl, benzoylacetyl, methylphenylglyoxal, pyruvophenone, 1,2-propanedione, 1-phenyl, phenylmethyldiketone, benzoyl methyl ketone, methyl phenyl diketone
CHEBI:63552
CC(=O)C(=O)C1=CC=CC=C1

Specifications

579-07-7
100.0
1.1000g/mL
84°C
97.5% min. (GC)
C9H8O2
C6H5COCOCH3
1 g
1.1
Solubility in water: insoluble
CC(=O)C(=O)C1=CC=CC=C1
148.16
CHEBI:63552
98%
1-Phenyl-1, 2-propanedione, 98%
97.5
Yellow
103.0°C to 105.0°C (14.0 mmHg)
Authentic
Glass bottle
1.5300 to 1.5340
MFCD00008755
07, 677
1-phenyl-1,2-propanedione, acetylbenzoyl, acetyl benzoyl, benzoylacetyl, methylphenylglyoxal, pyruvophenone, 1,2-propanedione, 1-phenyl, phenylmethyldiketone, benzoyl methyl ketone, methyl phenyl diketone
BVQVLAIMHVDZEL-UHFFFAOYSA-N
1-phenylpropane-1,2-dione
11363
148.16
Liquid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 209-435-2

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only