missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenyl-2-thiourea, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 207250250

 View more versions of this product

Catalog No. AC207250250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Phenyl-2-thiourea
96.0
148°C to 150°C
Authentic
Glass bottle
C6H5NHCSNH2
25 g
15, 7424
Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol,aq.ether,naoh,ethanol
NC(=S)NC1=CC=CC=C1
152.22
CHEBI:46261
97%
103-85-5
100.0
White to Beige
96% min. (HPLC)
C7H8N2S
MFCD00004933
12, 388
n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
FULZLIGZKMKICU-UHFFFAOYSA-N
phenylthiourea
676454
152.21
Crystalline Powder

Chemical Identifiers

103-85-5
152.22
FULZLIGZKMKICU-UHFFFAOYSA-N
676454
phenylthiourea
C7H8N2S
MFCD00004933
n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
CHEBI:46261
NC(=S)NC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Fatal if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTE

GHS Signal Word: Danger

EINECSNumber : 203-151-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only