Learn More
1-Phenyl-2-thiourea, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 207251000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Phenyl-2-thiourea | |
96.0 | |
148°C to 150°C | |
Authentic | |
Glass bottle | |
C6H5NHCSNH2 | |
100 g | |
15, 7424 | |
Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol,aq.ether,naoh,ethanol | |
NC(=S)NC1=CC=CC=C1 | |
152.22 | |
CHEBI:46261 | |
97% |
103-85-5 | |
100.0 | |
White to Beige | |
96% min. (HPLC) | |
C7H8N2S | |
MFCD00004933 | |
12, 388 | |
n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea | |
FULZLIGZKMKICU-UHFFFAOYSA-N | |
phenylthiourea | |
676454 | |
152.21 | |
Crystalline Powder |
Chemical Identifiers
103-85-5 | |
152.22 | |
FULZLIGZKMKICU-UHFFFAOYSA-N | |
676454 | |
phenylthiourea |
C7H8N2S | |
MFCD00004933 | |
n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea | |
CHEBI:46261 | |
NC(=S)NC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Fatal if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTE
GHS Signal Word: Danger
EINECSNumber : 203-151-2
RUO – Research Use Only