missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(Phenylsulfonyl)-1H-indole-2-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC03301DA

 View more versions of this product

Catalog No. CC03301DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

1-(Phenylsulfonyl)-1H-indole-2-carboxylic acid
96.5
97%
C15H11NO4S
1g
QIWDUGKJAHJRAE-UHFFFAOYSA-N
1-(benzenesulfonyl)indole-2-carboxylic acid
2776219
97%
40899-93-2
100.0
Amber glass bottle
MFCD03086089
1-phenylsulfonyl-1h-indole-2-carboxylic acid, 1-benzenesulfonyl indole-2-carboxylic acid, 1-benzenesulfonyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylicacid, 1-phenylsulfonyl
C1=CC=C(C=C1)S(=O)(=O)N2C3=CC=CC=C3C=C2C(=O)O
301.316
301.32

Chemical Identifiers

40899-93-2
301.316
QIWDUGKJAHJRAE-UHFFFAOYSA-N
2776219
C1=CC=C(C=C1)S(=O)(=O)N2C3=CC=CC=C3C=C2C(=O)O
C15H11NO4S
MFCD03086089
1-phenylsulfonyl-1h-indole-2-carboxylic acid, 1-benzenesulfonyl indole-2-carboxylic acid, 1-benzenesulfonyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylicacid, 1-phenylsulfonyl
1-(benzenesulfonyl)indole-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions