missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propanesulfonyl chloride, 97%, Thermo Scientific Chemicals

$91.04 - $252.45

Chemical Identifiers

CAS 10147-36-1
Molecular Formula C3H7ClO2S
Molecular Weight (g/mol) 142.597
MDL Number MFCD00007462
InChI Key KPBSJEBFALFJTO-UHFFFAOYSA-N
Synonym 1-propanesulfonyl chloride, propanesulfonyl chloride, propanesulphonyl chloride, propylsulfonyl chloride, 1-propanesulfonylchloride, n-propanesulfonyl chloride, 1-propane sulfonyl chloride, chloropropylsulfone, n-prso2cl, propanesulfonylchloride
PubChem CID 66279
IUPAC Name propane-1-sulfonyl chloride
SMILES CCCS(=O)(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60747AC
View Documents
Thermo Scientific Chemicals
H60747AC
25 mL
Each for $91.04
 
AAH60747AE
View Documents
Thermo Scientific Chemicals
H60747AE
100 mL
Each for $252.45
 
Description

Description

1-Propanesulfonyl chloride was used in synthesis of 1-alkyl-3-methylimidazolium alkanesulfonate ionic liquids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Propanesulfonyl chloride was used in synthesis of 1-alkyl-3-methylimidazolium alkanesulfonate ionic liquids.

Solubility
Decomposes in hot water.

Notes
Moisture sensitive. Store in dry temperature.
Specifications

Chemical Identifiers

10147-36-1
142.597
KPBSJEBFALFJTO-UHFFFAOYSA-N
66279
CCCS(=O)(=O)Cl
C3H7ClO2S
MFCD00007462
1-propanesulfonyl chloride, propanesulfonyl chloride, propanesulphonyl chloride, propylsulfonyl chloride, 1-propanesulfonylchloride, n-propanesulfonyl chloride, 1-propane sulfonyl chloride, chloropropylsulfone, n-prso2cl, propanesulfonylchloride
propane-1-sulfonyl chloride

Specifications

10147-36-1
78°C to 79°C
Pungent, Makes Eyes Water
1.453
MFCD00007462
UN3265
Moisture sensitive
Decomposes in hot water.
CCCS(=O)(=O)Cl
142.597
142.6
1-Propanesulfonyl chloride
1.267
97°C (207°F)
C3H7ClO2S
CH3CH2CH2SO2Cl
25 mL
1747500
1-propanesulfonyl chloride, propanesulfonyl chloride, propanesulphonyl chloride, propylsulfonyl chloride, 1-propanesulfonylchloride, n-propanesulfonyl chloride, 1-propane sulfonyl chloride, chloropropylsulfone, n-prso2cl, propanesulfonylchloride
KPBSJEBFALFJTO-UHFFFAOYSA-N
propane-1-sulfonyl chloride
66279
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 233-414-7

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only