missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propanol, 99.5%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 396940010

 View more versions of this product

Catalog No. AC396940010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Propanol
Extra Dry
99.4
0.8000g/mL
1 L
Authentic
1.3840 to 1.3860
99.5%
C3H8O
MFCD00002941
0.005% Max. (Coulometric)
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.1
Extra Dry
-127.0°C
71-23-8
100.0
97.0°C
15°C
Liquid
0.001 % max.
0.8
CH3CH2CH2OH
2.2 mPa.s (20°C)
15, 7955
Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents
CCCO
60.10
CHEBI:28831
99.5%

Chemical Identifiers

71-23-8
60.10
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CHEBI:28831
CCCO
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only