Learn More
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
$192.56 - $1061.00
Chemical Identifiers
CAS | 71-23-8 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.10 |
MDL Number | MFCD00002941 |
InChI Key | BDERNNFJNOPAEC-UHFFFAOYSA-N |
Synonym | 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol |
PubChem CID | 1031 |
ChEBI | CHEBI:28831 |
IUPAC Name | propan-1-ol |
SMILES | CCCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC149480025
|
Thermo Scientific Chemicals
149480025 |
2.5 L | Glass Bottle |
Each for $192.56
Case of 4 Each for $0.00
|
|
||||
AC149480250
|
Thermo Scientific Chemicals
149480250 |
25 L | Metal Drum |
Each for $1,061.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
71-23-8 | |
60.10 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
1031 | |
propan-1-ol |
C3H8O | |
MFCD00002941 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CHEBI:28831 | |
CCCO |
Specifications
71-23-8 | |
100.0 | |
2.5 L | |
Glass Bottle | |
1.3840 to 1.3860 (20°C, 589nm) | |
99+% | |
CH3CH2CH2OH | |
Vapor Pressure: 25mbar at 20°C | |
01, IV, 1413 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CCCO | |
60.10 | |
CHEBI:28831 | |
≥99.0% | |
1-Propanol |
99.0 | |
0.8000g/mL | |
Authentic | |
Liquid | |
0.005% max. | |
C3H8O | |
MFCD00002941 | |
0.5% max. | |
15, 7955 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
propan-1-ol | |
1031 | |
60.1 | |
Extra Pure |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye damage.
Highly flammable liquid and vapor.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only