missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 041842AK

 View more versions of this product

Catalog No. AA41842AK


Explore available promotions

Description

Description

Used in making cosmetics, skin and hair preparations, pharmaceuticals, perfumes, lacquer formulations, dye solutions, antifreezes, rubbing alcohols, soaps, window cleaners, acetone and other chemicals and products. Also referred as ropyl alcohol;n-Propan-1-ol;n-Propanol;n-Propyl alcohol, Ethylcarbinol, Optal, Osmosol extra.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Propanol
71-23-8
Clear liquid
97°C
15°C (59°F)
Alcohol-like
1.385
C3H8O
13.3 hPa (10mm Hg) at 20°C
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.1
°C to 127°C
Colorless
0.804
250 mL
824°C (1515°F)
Liquid
99.9%
MFCD00002941
1098242
Completely soluble in water. Miscible with propylene glycol.
CCCO
60.10
CHEBI:28831

Chemical Identifiers

71-23-8
60.10
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CHEBI:28831
CCCO
Videos
Safety and Handling

Safety and Handling

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P370+P378q-P501c

H225-H318-H336

DOTInformation : Hazard Class: 3; Packaging Group: II

EINECSNumber : 200-746-9

RTECSNumber : UH8225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only