missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propanol, anhydrous, 99.9%, Thermo Scientific Chemicals

Water <100ppm

Supplier:  Thermo Scientific Chemicals 041465K7

 View more versions of this product

Catalog No. AA41465K7


Explore available promotions

Description

Description

Solvent1-Propanol is used as a solvent and an entrainer in azeotropic distillation. It is used as a precursor to prepare propyl acetate, propylamide and propyl halides by reacting with acetic acid, ammonia and halogens respectively. It is actively used as a solvent in inks for flexographic printing, which is a technique used for printing. It is added to cleaners, floor polishes and metal degreasing fluids and is used as an additional solvent in the manufacture of adhesives. It enhances the drying characteristics of alkyd resins, electrodeposition paints and baking finishes in coating industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Propanol
71-23-8
Clear liquid
97°C
15°C (59°F)
Alcohol-like
1.385
C3H8O
13.3 hPa (10mm Hg) at 20°C
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.1
-127°C
Colorless
0.804
4 L
824°C (1515°F)
Liquid
99.9%
MFCD00002941
1098242
Miscible with water,alcohols,ketones,aldehydes,ethers,glycols,aromatic and aliphatic hydrocarbons.
CCCO
60.10
CHEBI:28831

Chemical Identifiers

71-23-8
60.10
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CHEBI:28831
CCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P370+P378q-P501c

H225-H318-H336

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 200-746-9

RTECSNumber : UH8225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only