missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propanol (Certified), Fisher Chemical™

Supplier:  Thermo Fisher Scientific BPA4141

 View more versions of this product

Catalog No. A414-1



Explore available promotions

Description

Description

Specifications

Specifications

7
-127°C
Colorless
0.800g/cm³
1 L
Glass Bottle
0.01% max.
C3H8O
MFCD00002941
2.2 mPa/s at 20°C
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.09
1-Propanol
71-23-8
0.02% max.
97°C
Pass Test
Liquid
0.801 to 0.803 (at 25°C)
CH3CH2CH2OH
25mbar at 20°C
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CCCO
60.10
CHEBI:28831
Certified

Chemical Identifiers

71-23-8
60.10
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CHEBI:28831
CCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. May cause skin and respiratory tract irritation. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Promotions

Promotions