missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Propanol, CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haen
CHROMASOLV™, for HPLC,≥99.9%
$294.58 - $3937.13
Chemical Identifiers
CAS | 71-23-8 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.10 |
MDL Number | MFCD00002941 |
InChI Key | BDERNNFJNOPAEC-UHFFFAOYSA-N |
PubChem CID | 1031 |
ChEBI | CHEBI:28831 |
IUPAC Name | propan-1-ol |
SMILES | CCCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6000099
|
Honeywell Chemicals
348711L |
1 L | Glass Bottle |
Each for $294.58
|
|
||||
6000100
|
Honeywell Chemicals
3487125L |
2.5 L | Glass Bottle |
Each for $531.53
|
|
||||
60046493
|
Honeywell Chemicals
348716X1L |
6 x 1 L | Glass Bottle |
Case of 6 Each for $1,433.08
|
|
||||
6000102
|
Honeywell Chemicals
348714X25L |
4 x 2.5 L | Glass Bottle |
Case of 4 Each for $1,885.48
|
|
||||
60048244
|
Honeywell Chemicals
348714X4L |
4 x 4 L | Glass Bottle |
Case of 4 Each for $3,937.13
|
|
||||
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Chemical Identifiers
71-23-8 | |
60.10 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
CHEBI:28831 | |
CCCO |
C3H8O | |
MFCD00002941 | |
1031 | |
propan-1-ol |
Specifications
-127°C (literature) | |
100.0000% | |
96°C to 98°C | |
21°C | |
C3H8O | |
MFCD00002941 | |
1098242 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
propan-1-ol | |
1031 | |
60.1 | |
HPLC |
71-23-8 | |
0.804Â g/mLÂ at 25Â °C(lit.) | |
1 L | |
Glass Bottle | |
CH3CH2CH2OH | |
UN1274 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CCCO | |
60.10 | |
CHEBI:28831 | |
>99.9% | |
1-Propanol, CHROMASOLV™, for HPLC, ≥99.9% |
Safety and Handling
ShelfLife : 1620 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: n-Propanol
EINECSNumber : 200-746-9
Recommended Storage : Room Temp