Learn More
1-Propanol, for HPLC
CAS: 71-23-8 | C3H8O | 60.10 g/mol
$240.73 - $430.43
Chemical Identifiers
| CAS | 71-23-8 |
|---|---|
| Molecular Formula | C3H8O |
| Molecular Weight (g/mol) | 60.10 |
| MDL Number | MFCD00002941 |
| InChI Key | BDERNNFJNOPAEC-UHFFFAOYSA-N |
| Synonym | 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol |
| PubChem CID | 1031 |
| ChEBI | CHEBI:28831 |
| IUPAC Name | propan-1-ol |
| SMILES | CCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC389600010
|
Thermo Scientific Chemicals
389600010 |
1 L | Glass bottle |
Each for $240.73
|
|
||||
|
AC389600025
|
Thermo Scientific Chemicals
389600025 |
2.5 L | Glass bottle |
Each for $430.43
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLCChemical Identifiers
| 71-23-8 | |
| 60.10 | |
| BDERNNFJNOPAEC-UHFFFAOYSA-N | |
| 1031 | |
| propan-1-ol |
| C3H8O | |
| MFCD00002941 | |
| 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
| CHEBI:28831 | |
| CCCO |
Specifications
| -127.0°C | |
| 0.05 max. (1 cm cell vs HPLC−grade water) at 250nm, 0.3 max. (1 cm cell vs HPLC−grade water) at 220nm, 0.7 max. (1 cm cell vs HPLC−grade water) at 210nm | |
| 0.8000g/mL | |
| 1 L | |
| Glass bottle | |
| 1.3840 to 1.3860 | |
| C3H8O | |
| MFCD00002941 | |
| 15, 7955 | |
| Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents | |
| CCCO | |
| 60.10 | |
| CHEBI:28831 | |
| ≥99.8% | |
| 1-Propanol |
| 71-23-8 | |
| Colorless | |
| 97.0°C | |
| 15°C | |
| Liquid | |
| 0.8 | |
| CH3CH2CH2OH | |
| 2.2 mPa.s (20°C) | |
| 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
| BDERNNFJNOPAEC-UHFFFAOYSA-N | |
| propan-1-ol | |
| 1031 | |
| 60.1 | |
| HPLC |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Causes serious eye damage.
Highly flammable liquid and vapor.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
EINECSNumber : 200-746-9
RUO – Research Use Only