missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propanol, puriss. p.a., ACS Reagent, ≥99.5% (GC), Honeywell™ Riedel-de Haën™

Puriss. p.a., Reag. Ph. Eur.,≥99.5% (GC)

$353.10 - $1765.30

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046266
View Documents
Honeywell-Riedel-de Haen
335381L
1 L Poly Bottle
Each for $353.10
 
60046267
View Documents
Honeywell-Riedel-de Haen
3353825L
2.5 L Poly Bottle
Each for $793.35
 
60046269
View Documents
Honeywell-Riedel-de Haen
335386X1L
6 x 1 L Poly Bottle
Case of 6 Each for $1,765.30
 
60046268
View Documents
Honeywell-Riedel-de Haen
335384X25L
4 x 2.5 L Poly Bottle
Case of 4 Each for $585.48
 
Description

Description

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications

Chemical Identifiers

71-23-8
60.10
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CHEBI:28831
CCCO

Specifications

-127°C
Colorless
97°C
0.137
700°F
Poly Bottle
C3H8O
MFCD00002941
UN1274
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.1g/mol
1-Propanol
71-23-8
0.804g/mL at 25°C
1 L
22°C (71.6°F)
Alcohol-like
n20/D 1.384
CH3CH2CH2OH
14.9mmHg (20°C), 10mmHg (147°C)
1098242
Soluble in water and most organic solvents
CCCO
60.10
CHEBI:28831
≥99.5% (GC)
Videos
Safety and Handling

Safety and Handling

P210-P261-P280-P305 + P351 + P338

DOTInformation : Class 3 - PG 2 - n-Propanol

RTECSNumber : 200-746-9

SDS
missing translation for 'documents'

missing translation for 'documents'