missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propanol, purum, ≥99% (GC), Honeywell™

≥99% (GC), Purum

Supplier:  Honeywell Chemicals 241356X1L

 View more versions of this product

Catalog No. 60045526


Explore available promotions

Description

Description

  • ≥95% for general laboratory use.
Specifications

Specifications

1-Propanol
71-23-8
97°
0.137
700°F
Liquid
CH3CH2CH2OH
10mmHg (147°C), 14.9mmHg (20°C)
1098242
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.1g/mol
purum
-127°
0.804g/mL at 25°C
6 x 1 L
22°C (71.6°F)
Poly Bottle
C3H8O
MFCD00002941
UN1274
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CCCO
60.10
CHEBI:28831
≥99% (GC)

Chemical Identifiers

71-23-8
60.10
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CHEBI:28831
CCCO
Videos
Safety and Handling

Safety and Handling

P210-P261-P280-P305 + P351 + P338

H225-H318-H336

DOTInformation : Class 3 - PG 2 - n-Propanol

EINECSNumber : 200-746-9

RTECSNumber : UH8225000

SDS
Documents

Documents

Promotions

Promotions