missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propene 1,3-Sultone 99.0+%, TCI America™

Supplier:  TCI America P226025G

 View more versions of this product

Catalog No. P226025G


Explore available promotions

Description

Description

Specifications

Specifications

1-Propene 1,3-Sultone
85°C
118°C
MFCD12405143
5H-1,2-Oxathiole 2,2-Dioxide
C1C=CS(=O)(=O)O1
120.122
120.12
Crystalline Powder
21806-61-1
White
C3H4O3S
25 g
KLLQVNFCMHPYGL-UHFFFAOYSA-N
5H-oxathiole 2,2-dioxide
10898703
≥99.0% (GC)

Chemical Identifiers

21806-61-1
120.122
KLLQVNFCMHPYGL-UHFFFAOYSA-N
10898703
C1C=CS(=O)(=O)O1
C3H4O3S
MFCD12405143
5H-1,2-Oxathiole 2,2-Dioxide
5H-oxathiole 2,2-dioxide
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions