missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Propene 1,3-Sultone 99.0+%, TCI America™
Supplier: TCI America P22605G
Specifications
1-Propene 1,3-Sultone | |
85°C | |
118°C | |
MFCD12405143 | |
5H-1,2-Oxathiole 2,2-Dioxide | |
C1C=CS(=O)(=O)O1 | |
120.122 | |
120.12 | |
Crystalline Powder |
21806-61-1 | |
White | |
C3H4O3S | |
5 g | |
KLLQVNFCMHPYGL-UHFFFAOYSA-N | |
5H-oxathiole 2,2-dioxide | |
10898703 | |
≥99.0% (GC) |
Chemical Identifiers
21806-61-1 | |
120.122 | |
KLLQVNFCMHPYGL-UHFFFAOYSA-N | |
10898703 | |
C1C=CS(=O)(=O)O1 |
C3H4O3S | |
MFCD12405143 | |
5H-1,2-Oxathiole 2,2-Dioxide | |
5H-oxathiole 2,2-dioxide |
Safety and Handling
TSCA : No