missing translation for 'onlineSavingsMsg'
Learn More

1-Propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

CAS: 14092-04-7 | C3H5BrMg | 145.28 g/mol

$252.11 - $252.11

Chemical Identifiers

CAS 14092-04-7
Molecular Formula C3H5BrMg
Molecular Weight (g/mol) 145.28
MDL Number MFCD00209610
InChI Key HHNLJRLNDSEYOX-UHFFFAOYSA-M
Synonym 1-propenylmagnesium bromide, grignard reagent, propenylmagnesium bromide, magnesium, bromo-1z-1-propenyl, magnesium, bromo-1z-1-propenyl, e-prop-1-en-1-ylmagnesium bromide, propenyl magnesium bromide, prop-1-enylmagnesium bromide, prop-1-enylmagnesium bromide, prop-1-en-1-ylmagnesium bromide, prop-1-en-1-ylmagnesium bromide
PubChem CID 5112801
IUPAC Name magnesium;prop-1-ene;bromide
SMILES C\C=C\[Mg]Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC434671000
View Documents
Thermo Scientific Chemicals
434671000
100 mL AcroSeal™ Glass Bottle
Each for $252.11
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14092-04-7
145.28
HHNLJRLNDSEYOX-UHFFFAOYSA-M
5112801
C\C=C\[Mg]Br
C3H5BrMg
MFCD00209610
1-propenylmagnesium bromide, grignard reagent, propenylmagnesium bromide, magnesium, bromo-1z-1-propenyl, magnesium, bromo-1z-1-propenyl, e-prop-1-en-1-ylmagnesium bromide, propenyl magnesium bromide, prop-1-enylmagnesium bromide, prop-1-enylmagnesium bromide, prop-1-en-1-ylmagnesium bromide, prop-1-en-1-ylmagnesium bromide
magnesium;prop-1-ene;bromide

Specifications

14092-04-7,109-99-9
8.41,93.12
0.9500g/mL
−20°C
C3H5BrMg
100 mL
1-propenylmagnesium bromide, grignard reagent, propenylmagnesium bromide, magnesium, bromo-1z-1-propenyl, magnesium, bromo-1z-1-propenyl, e-prop-1-en-1-ylmagnesium bromide, propenyl magnesium bromide, prop-1-enylmagnesium bromide, prop-1-enylmagnesium bromide, prop-1-en-1-ylmagnesium bromide, prop-1-en-1-ylmagnesium bromide
HHNLJRLNDSEYOX-UHFFFAOYSA-M
magnesium;prop-1-ene;bromide
5112801
Solution
6.88,91.59
Brown to Yellow
65.0°C
AcroSeal™ Glass Bottle
MFCD00209610
0.95
Solubility in water: reacts.
C\C=C\[Mg]Br
145.28
145.28
1-Propenylmagnesium bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently w

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only