missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propoxy-2-propanol (contains 2-Isopropoxy-1-propanol) 93.0+%, TCI America™

Supplier:  TCI America P2042500ML

 View more versions of this product

Catalog No. P2042500ML


Explore available promotions

Description

Description

Specifications

Specifications

1-Propoxy-2-propanol (contains 2-Isopropoxy-1-propanol)
Yellow
C6H14O2
500 mL
Propylene Glycol Monopropyl Ether
CCCOCC(C)O
118.18
118.18
Liquid
1569-01-3
149°C
MFCD00192420
1993
FENFUOGYJVOCRY-UHFFFAOYNA-N
1-propoxypropan-2-ol
15286
≥93.0% (GC)

Chemical Identifiers

1569-01-3
118.18
FENFUOGYJVOCRY-UHFFFAOYNA-N
15286
CCCOCC(C)O
C6H14O2
MFCD00192420
Propylene Glycol Monopropyl Ether
1-propoxypropan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2424&(7)-0097

RTECSNumber : UB9351000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions