missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1-Propyl)triphenylphosphonium bromide, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1296830

 View more versions of this product

Catalog No. AAA1296830


Explore available promotions

Description

Description

It is employed as a catalyst for the carboxylation of styrene oxide and as a reactant involved in epoxidation of arylalkenes, boration of allylic epoxides, Wittig reactions, selective dimerization of α-olefins, and synthesis of indoles. It has also been shown to be part of Phosphonium salts, having antiviral activity against influenza virus A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a catalyst for the carboxylation of styrene oxide and as a reactant involved in epoxidation of arylalkenes, boration of allylic epoxides, Wittig reactions, selective dimerization of α-olefins, and synthesis of indoles. It has also been shown to be part of Phosphonium salts, having antiviral activity against influenza virus A.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, humidity and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Specifications

(1-Propyl)triphenylphosphonium bromide
236°C to 240°C
MFCD00011843
3599726
propyltriphenylphosphonium bromide, triphenyl propyl phosphonium bromide, 1-propyl triphenylphosphonium bromide, n-propyltriphenylphosphonium bromide, triphenyl propyl phosphanium bromide, n-propyl triphenylphosphonium bromide, propyl triphenyl phosphonium chloride, acmc-1bff2, propyltriphenylphosphoniumbromide
XMQSELBBYSAURN-UHFFFAOYSA-M
triphenyl(propyl)phosphanium;bromide
80374
99%
6228-47-3
C21H22BrP
250 g
Hygroscopic
Soluble in water.
[Br-].CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
385.29
385.29

Chemical Identifiers

6228-47-3
385.29
XMQSELBBYSAURN-UHFFFAOYSA-M
80374
[Br-].CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C21H22BrP
MFCD00011843
propyltriphenylphosphonium bromide, triphenyl propyl phosphonium bromide, 1-propyl triphenylphosphonium bromide, n-propyltriphenylphosphonium bromide, triphenyl propyl phosphanium bromide, n-propyl triphenylphosphonium bromide, propyl triphenyl phosphonium chloride, acmc-1bff2, propyltriphenylphosphoniumbromide
triphenyl(propyl)phosphanium;bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 228-330-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only