missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-S-Octyl-β-D-thioglucopyranoside, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 85618-21-9 |
---|---|
Molecular Formula | C14H28O5S |
Molecular Weight (g/mol) | 308.44 |
MDL Number | MFCD00012189 |
InChI Key | CGVLVOOFCGWBCS-DGTMBMJNSA-N |
Synonym | octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside |
PubChem CID | 53720324 |
IUPAC Name | (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol |
SMILES | CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC315000025
|
Thermo Scientific Chemicals
315000025 |
2.5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC315000100
|
Thermo Scientific Chemicals
315000100 |
10g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
85618-21-9 | |
308.44 | |
CGVLVOOFCGWBCS-DGTMBMJNSA-N | |
53720324 | |
CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O |
C14H28O5S | |
MFCD00012189 | |
octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside | |
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol |
Specifications
85618-21-9 | |
100.0 | |
97.5% min. (ELSD) (HPLC) | |
C14H28O5S | |
2.5g | |
-46 | |
Solubility in water: soluble | |
CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O | |
308.44 | |
308.44 | |
1-S-Octyl-β-D-thioglucopyranoside, 98% |
97.5 | |
Authentic | |
Glass bottle | |
MFCD00012189 | |
− 46.00 (20.00°C c=1,CH3OH) | |
octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside | |
CGVLVOOFCGWBCS-DGTMBMJNSA-N | |
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol | |
53720324 | |
98% |
Safety and Handling
TSCA : TSCA
RUO â Research Use Only