missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-tert-Butyl N-(tert-Butoxycarbonyl)-L-aspartate 95.0+%, TCI America™
$85.75 - $206.90
Chemical Identifiers
| CAS | 34582-32-6 |
|---|---|
| Molecular Formula | C13H23NO6 |
| Molecular Weight (g/mol) | 289.328 |
| MDL Number | MFCD00038272 |
| InChI Key | RAUQRYTYJIYLTF-QMMMGPOBSA-N |
| Synonym | boc-asp-otbu, boc-l-aspartic acid 1-tert-butyl ester, s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-tert-boc-l-aspartic acid tert-butyl ester, 3s-4-tert-butoxy-3-tert-butoxycarbonylamino-4-oxo-butanoic acid, 3s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp-otbu, boc-asp-o-t-butyl, pubchem12140, boc-aspartic acid-otbu |
| PubChem CID | 7010517 |
| IUPAC Name | (3S)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid |
| SMILES | CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C |
Chemical Identifiers
| 34582-32-6 | |
| 289.328 | |
| RAUQRYTYJIYLTF-QMMMGPOBSA-N | |
| 7010517 | |
| CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C |
| C13H23NO6 | |
| MFCD00038272 | |
| boc-asp-otbu, boc-l-aspartic acid 1-tert-butyl ester, s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-tert-boc-l-aspartic acid tert-butyl ester, 3s-4-tert-butoxy-3-tert-butoxycarbonylamino-4-oxo-butanoic acid, 3s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp-otbu, boc-asp-o-t-butyl, pubchem12140, boc-aspartic acid-otbu | |
| (3S)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid |
Specifications
| 34582-32-6 | |
| White | |
| MFCD00038272 | |
| boc-asp-otbu, boc-l-aspartic acid 1-tert-butyl ester, s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-tert-boc-l-aspartic acid tert-butyl ester, 3s-4-tert-butoxy-3-tert-butoxycarbonylamino-4-oxo-butanoic acid, 3s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp-otbu, boc-asp-o-t-butyl, pubchem12140, boc-aspartic acid-otbu | |
| CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C | |
| 289.328 | |
| 289.33 | |
| Crystalline Powder |
| 103°C | |
| C13H23NO6 | |
| 1 g | |
| RAUQRYTYJIYLTF-QMMMGPOBSA-N | |
| (3S)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid | |
| 7010517 | |
| ≥95.0% (HPLC,T) | |
| 1-tert-Butyl N-(tert-Butoxycarbonyl)-L-aspartate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator