missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-tert-Butylimidazole 98.0+%, TCI America™
$514.48 - $1534.11
Chemical Identifiers
| CAS | 45676-04-8 |
|---|---|
| Molecular Formula | C7H12N2 |
| Molecular Weight (g/mol) | 124.19 |
| MDL Number | MFCD19441219 |
| InChI Key | AMQKPABOPFXDQM-UHFFFAOYSA-N |
| PubChem CID | 142665 |
| IUPAC Name | 1-tert-butyl-1H-imidazole |
| SMILES | CC(C)(C)N1C=CN=C1 |
Chemical Identifiers
| 45676-04-8 | |
| 124.19 | |
| AMQKPABOPFXDQM-UHFFFAOYSA-N | |
| 1-tert-butyl-1H-imidazole |
| C7H12N2 | |
| MFCD19441219 | |
| 142665 | |
| CC(C)(C)N1C=CN=C1 |
Specifications
| 45676-04-8 | |
| 105°C | |
| MFCD19441219 | |
| AMQKPABOPFXDQM-UHFFFAOYSA-N | |
| 1-tert-butyl-1H-imidazole | |
| 142665 | |
| ≥98.0% (GC,T) | |
| 1-tert-Butylimidazole |
| Yellow | |
| C7H12N2 | |
| 5 g | |
| CC(C)(C)N1C=CN=C1 | |
| 124.19 | |
| 124.19 | |
| Liquid |
Safety and Handling
TSCA : No