Learn More
1-Tetradecyne, tech. 90%, Thermo Scientific Chemicals
$113.30 - $381.97
Chemical Identifiers
CAS | 765-10-6 |
---|---|
Molecular Formula | C14H26 |
Molecular Weight (g/mol) | 194.362 |
MDL Number | MFCD00015076 |
InChI Key | DZEFNRWGWQDGTR-UHFFFAOYSA-N |
Synonym | 1-tetradecyne, tetradecyne, tetradec-1-in, acmc-1bfxn |
PubChem CID | 69823 |
IUPAC Name | tetradec-1-yne |
SMILES | CCCCCCCCCCCCC#C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0257306
|
Thermo Scientific Chemicals
L0257306 |
5 g |
Each for $113.30
|
|
|||||
AAL0257314
|
Thermo Scientific Chemicals
L0257314 |
25 g |
Each for $381.97
|
|
|||||
Description
1-Tetradecyne, is used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Tetradecyne, is used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.
Solubility
Not miscible with water. soluble in common organic solvents of low polarity
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Chemical Identifiers
765-10-6 | |
194.362 | |
DZEFNRWGWQDGTR-UHFFFAOYSA-N | |
69823 | |
CCCCCCCCCCCCC#C |
C14H26 | |
MFCD00015076 | |
1-tetradecyne, tetradecyne, tetradec-1-in, acmc-1bfxn | |
tetradec-1-yne |
Specifications
765-10-6 | |
132°C to 134°C (14 mmHg) | |
1.441 | |
5 g | |
1-tetradecyne, tetradecyne, tetradec-1-in, acmc-1bfxn | |
DZEFNRWGWQDGTR-UHFFFAOYSA-N | |
tetradec-1-yne | |
69823 | |
90% | |
1-Tetradecyne |
0.79 | |
C14H26 | |
MFCD00015076 | |
1746546 | |
Not miscible with water. soluble in common organic solvents of low polarity | |
CCCCCCCCCCCCC#C | |
194.362 | |
194.36 | |
Technical |
Safety and Handling
EINECSNumber : 212-139-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only