Learn More
1-Trifluoromethylcyclopropane-1-carboxylic acid, 97%, Thermo Scientific Chemicals
$468.41 - $468.41
Chemical Identifiers
CAS | 277756-46-4 |
---|---|
Molecular Formula | C5H5F3O2 |
Molecular Weight (g/mol) | 154.088 |
MDL Number | MFCD03093070 |
InChI Key | SKCBKBCACWDALV-UHFFFAOYSA-N |
Synonym | 1-trifluoromethyl cyclopropane-1-carboxylic acid, 1-trifluoromethyl cyclopropanecarboxylic acid, 1-trifluoromethylcyclopropanecarboxylic acid, 1-trifluoromethyl cyclopropane-1-carboxylicacid, 1-trifluoromethyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-trifluoromethyl, 1-trifluoromethyl-cyclopropanecarboxylic acid, pubchem20086, acmc-20aa6t, 1-trifluoromethylcyclopropanecarboxylicacid |
PubChem CID | 2778306 |
IUPAC Name | 1-(trifluoromethyl)cyclopropane-1-carboxylic acid |
SMILES | C1CC1(C(=O)O)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1795703
|
Thermo Scientific Chemicals
L1795703 |
1 g |
Each for $468.41
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
277756-46-4 | |
154.088 | |
SKCBKBCACWDALV-UHFFFAOYSA-N | |
2778306 | |
C1CC1(C(=O)O)C(F)(F)F |
C5H5F3O2 | |
MFCD03093070 | |
1-trifluoromethyl cyclopropane-1-carboxylic acid, 1-trifluoromethyl cyclopropanecarboxylic acid, 1-trifluoromethylcyclopropanecarboxylic acid, 1-trifluoromethyl cyclopropane-1-carboxylicacid, 1-trifluoromethyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-trifluoromethyl, 1-trifluoromethyl-cyclopropanecarboxylic acid, pubchem20086, acmc-20aa6t, 1-trifluoromethylcyclopropanecarboxylicacid | |
1-(trifluoromethyl)cyclopropane-1-carboxylic acid |
Specifications
277756-46-4 | |
182°C to 184°C | |
MFCD03093070 | |
UN3261 | |
1-trifluoromethyl cyclopropane-1-carboxylic acid, 1-trifluoromethyl cyclopropanecarboxylic acid, 1-trifluoromethylcyclopropanecarboxylic acid, 1-trifluoromethyl cyclopropane-1-carboxylicacid, 1-trifluoromethyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-trifluoromethyl, 1-trifluoromethyl-cyclopropanecarboxylic acid, pubchem20086, acmc-20aa6t, 1-trifluoromethylcyclopropanecarboxylicacid | |
C1CC1(C(=O)O)C(F)(F)F | |
154.088 | |
154.09 | |
1-Trifluoromethylcyclopropane-1-carboxylic acid |
87°C to 89°C | |
C5H5F3O2 | |
1 g | |
8543283 | |
SKCBKBCACWDALV-UHFFFAOYSA-N | |
1-(trifluoromethyl)cyclopropane-1-carboxylic acid | |
2778306 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only