missing translation for 'onlineSavingsMsg'
Learn More

1-Trimethylsilyl-1,4-pentadiyne, 98%

CAS: 71789-10-1 | C8H12Si | 136.27 g/mol

$58.26 - $782.03

Chemical Identifiers

CAS 71789-10-1
Molecular Formula C8H12Si
Molecular Weight (g/mol) 136.27
MDL Number MFCD00066350
InChI Key JKYMNPFNLMDSFG-UHFFFAOYSA-N
Synonym 1-trimethylsilyl-1,4-pentadiyne, trimethyl penta-1,4-diynyl silane, trimethyl penta-1,4-diyn-1-yl silane, 1,4-pentadiynyltrimethylsilane, silane,trimethyl-1,4-pentadiyn-1-yl
PubChem CID 144510
IUPAC Name trimethyl(penta-1,4-diynyl)silane
SMILES C[Si](C)(C)C#CCC#C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5343603
View Documents
Thermo Scientific Chemicals
H5343603
1 g
Each for $58.26
Only null left
 
AAH5343606
View Documents
Thermo Scientific Chemicals
H5343606
5 g
Each for $181.82
Only null left
 
AAH5343614
View Documents
Thermo Scientific Chemicals
H5343614
25 g
Each for $782.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with strong oxidizing agents and ammonia.
Specifications

Chemical Identifiers

71789-10-1
136.27
JKYMNPFNLMDSFG-UHFFFAOYSA-N
144510
C[Si](C)(C)C#CCC#C
C8H12Si
MFCD00066350
1-trimethylsilyl-1,4-pentadiyne, trimethyl penta-1,4-diynyl silane, trimethyl penta-1,4-diyn-1-yl silane, 1,4-pentadiynyltrimethylsilane, silane,trimethyl-1,4-pentadiyn-1-yl
trimethyl(penta-1,4-diynyl)silane

Specifications

42°C (25mmHg)
1 g
HC≡CCH2C≡CSi(CH3)3
UN1993
1-trimethylsilyl-1,4-pentadiyne, trimethyl penta-1,4-diynyl silane, trimethyl penta-1,4-diyn-1-yl silane, 1,4-pentadiynyltrimethylsilane, silane,trimethyl-1,4-pentadiyn-1-yl
C[Si](C)(C)C#CCC#C
136.27
136.27
0.789
71789-10-1
C8H12Si
MFCD00066350
8°C (46°F)
JKYMNPFNLMDSFG-UHFFFAOYSA-N
trimethyl(penta-1,4-diynyl)silane
144510
98%
1-Trimethylsilyl-1,4-pentadiyne
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only