missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

10-Bromo-1-decanol, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 384590010

 View more versions of this product

Catalog No. AC384590010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

10-Bromo-1-decanol
94.0
1.1900g/mL
109°C
94% min. (GC)
C10H21BrO
Br(CH2)10OH
1g
10-bromo-1-decanol, 10-bromodecanol, 1-decanol, 10-bromo, 10-hydroxy-1-bromo decane, 1-bromodecan-10-ol, 10-bromodecane-1-ol, 10-bromo-decan-1-ol, decamethylene bromohydrin, 10-hydroxy1-bromo decane, acmc-209l7p
OCCCCCCCCCCBr
237.18
237.18
53463-68-6
100.0
165°C to 169°C (10.0mmHg)
Authentic
Glass bottle
1.4780 to 1.4800 (20°C, 589nm)
MFCD00041681
1.19
LGZMUUBPTDRQQM-UHFFFAOYSA-N
10-bromodecan-1-ol
104507
95%

Chemical Identifiers

53463-68-6
237.18
LGZMUUBPTDRQQM-UHFFFAOYSA-N
104507
OCCCCCCCCCCBr
C10H21BrO
MFCD00041681
10-bromo-1-decanol, 10-bromodecanol, 1-decanol, 10-bromo, 10-hydroxy-1-bromo decane, 1-bromodecan-10-ol, 10-bromodecane-1-ol, 10-bromo-decan-1-ol, decamethylene bromohydrin, 10-hydroxy1-bromo decane, acmc-209l7p
10-bromodecan-1-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 258-572-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only