missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-Hydroxydecanoic acid, 96%
CAS: 1679-53-4 | C10H20O3 | 188.27 g/mol
$166.47 - $612.56
Chemical Identifiers
| CAS | 1679-53-4 |
|---|---|
| Molecular Formula | C10H20O3 |
| Molecular Weight (g/mol) | 188.27 |
| MDL Number | MFCD00010510 |
| InChI Key | YJCJVMMDTBEITC-UHFFFAOYSA-N |
| Synonym | 10-hydroxydecanoate, decanoic acid, 10-hydroxy, unii-np03xo416b, 10-hydroxy capric acid, 10-hydroxy-decanoic acid, 1-hydroxydecanoicacid, 10-oh-capric acid, 10-hydroxycapric acid, 10-oh-decanoic acid, 10-hydroxydecanoicacid |
| PubChem CID | 74300 |
| ChEBI | CHEBI:17409 |
| IUPAC Name | 10-hydroxydecanoic acid |
| SMILES | OCCCCCCCCCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6074303
|
Thermo Scientific Chemicals
H6074303 |
1 g |
N/A
|
|
|||||
|
AAH6074306
|
Thermo Scientific Chemicals
H6074306 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1679-53-4 | |
| 188.27 | |
| YJCJVMMDTBEITC-UHFFFAOYSA-N | |
| 74300 | |
| 10-hydroxydecanoic acid |
| C10H20O3 | |
| MFCD00010510 | |
| 10-hydroxydecanoate, decanoic acid, 10-hydroxy, unii-np03xo416b, 10-hydroxy capric acid, 10-hydroxy-decanoic acid, 1-hydroxydecanoicacid, 10-oh-capric acid, 10-hydroxycapric acid, 10-oh-decanoic acid, 10-hydroxydecanoicacid | |
| CHEBI:17409 | |
| OCCCCCCCCCC(O)=O |
Specifications
| 1679-53-4 | |
| C10H20O3 | |
| MFCD00010510 | |
| 10-hydroxydecanoate, decanoic acid, 10-hydroxy, unii-np03xo416b, 10-hydroxy capric acid, 10-hydroxy-decanoic acid, 1-hydroxydecanoicacid, 10-oh-capric acid, 10-hydroxycapric acid, 10-oh-decanoic acid, 10-hydroxydecanoicacid | |
| OCCCCCCCCCC(O)=O | |
| 188.27 | |
| CHEBI:17409 | |
| 96% |
| 75°C to 77°C | |
| HO(CH2)9CO2H | |
| 1 g | |
| YJCJVMMDTBEITC-UHFFFAOYSA-N | |
| 10-hydroxydecanoic acid | |
| 74300 | |
| 188.26 | |
| 10-Hydroxydecanoic acid |
Safety and Handling
EINECSNumber : 216-848-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only