missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

10-Methyl-9-phenylacridinium Perchlorate 98.0+%, TCI America™

$209.35 - $619.38

Chemical Identifiers

CAS 36519-61-6
Molecular Formula C20H16ClNO4
Molecular Weight (g/mol) 369.801
InChI Key NRNCXGCGIKATGE-UHFFFAOYSA-M
PubChem CID 11326154
IUPAC Name 10-methyl-9-phenylacridin-10-ium;perchlorate
SMILES C[N+]1=C2C=CC=CC2=C(C3=CC=CC=C31)C4=CC=CC=C4.[O-]Cl(=O)(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M17751G
View Documents
TCI America
M17751G
1 g
Each for $209.35
 
M17755G
View Documents
TCI America
M17755G
5 g
Each for $619.38
 
Description

Description

Specifications

Chemical Identifiers

36519-61-6
369.801
11326154
C[N+]1=C2C=CC=CC2=C(C3=CC=CC=C31)C4=CC=CC=C4.[O-]Cl(=O)(=O)=O
C20H16ClNO4
NRNCXGCGIKATGE-UHFFFAOYSA-M
10-methyl-9-phenylacridin-10-ium;perchlorate

Specifications

36519-61-6
1 g
NRNCXGCGIKATGE-UHFFFAOYSA-M
10-methyl-9-phenylacridin-10-ium;perchlorate
11326154
≥98.0% (HPLC,N)
10-Methyl-9-phenylacridinium Perchlorate
C20H16ClNO4
1479
C[N+]1=C2C=CC=CC2=C(C3=CC=CC=C31)C4=CC=CC=C4.[O-]Cl(=O)(=O)=O
369.801
369.80
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'