Biochrom 10 mm Quartz Matched Semi Micro Cuvette with Stopper
Accessory for use with Ultrospec 7000/8000/9000, Ultrospec 2100 pro, Novaspec III/Plus and GeneQuant 1300/100.
Supplier:  Biochrom 80210022
Catalog No. BC80210022
Explore available promotions
Description
Description
  • 10 mm
  • 800 μL working volume
  • Comes with stopper
  • Quartz
  • 2 Pk.
Specifications
Specifications
Cuvette with Stopper
Quartz
10 mm
800 μL
Stopper
Ultrospec 7000/8000/9000, Ultrospec 2100 Pro, Novaspec III/Plus and GeneQuant 1300/100 Spectrophotometers