Learn More
10-Phenylanthracene-9-boronic acid, 98%, Thermo Scientific Chemicals
$57.36 - $620.51
Chemical Identifiers
CAS | 334658-75-2 |
---|---|
Molecular Formula | C20H15BO2 |
Molecular Weight (g/mol) | 298.15 |
MDL Number | MFCD11111989 |
InChI Key | RVPCPPWNSMAZKR-UHFFFAOYSA-N |
Synonym | 10-phenylanthracen-9-yl boronic acid, 10-phenyl-9-anthracene boronic acid, 10-phenyl-9-anthraceneboronic acid, 10-phenylanthracene-9-boronic acid, 10-phenylantrhacen-9-yl boronic acid, boronic acid, 10-phenyl-9-anthracenyl, pubchem19639, 9-borono-10-phenylanthracene, 10-phenylanthracene-9-ylboronic acid, 10-phenyl-9-anthryl boronic acid |
PubChem CID | 22247164 |
IUPAC Name | (10-phenylanthracen-9-yl)boronic acid |
SMILES | OB(O)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH64911MD
|
Thermo Scientific Chemicals
H64911MD |
250 mg |
Each for $57.36
|
|
|||||
AAH6491103
|
Thermo Scientific Chemicals
H6491103 |
1 g |
Each for $159.85
|
|
|||||
AAH6491106
|
Thermo Scientific Chemicals
H6491106 |
5 g |
Each for $620.51
|
|
|||||
Description
It is used as pharmaceutical intermediate and as OLED materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediate and as OLED materials.
Solubility
Soluble in methanol.
Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Chemical Identifiers
334658-75-2 | |
298.15 | |
RVPCPPWNSMAZKR-UHFFFAOYSA-N | |
22247164 | |
OB(O)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12 |
C20H15BO2 | |
MFCD11111989 | |
10-phenylanthracen-9-yl boronic acid, 10-phenyl-9-anthracene boronic acid, 10-phenyl-9-anthraceneboronic acid, 10-phenylanthracene-9-boronic acid, 10-phenylantrhacen-9-yl boronic acid, boronic acid, 10-phenyl-9-anthracenyl, pubchem19639, 9-borono-10-phenylanthracene, 10-phenylanthracene-9-ylboronic acid, 10-phenyl-9-anthryl boronic acid | |
(10-phenylanthracen-9-yl)boronic acid |
Specifications
334658-75-2 | |
MFCD11111989 | |
11434760 | |
10-phenylanthracen-9-yl boronic acid, 10-phenyl-9-anthracene boronic acid, 10-phenyl-9-anthraceneboronic acid, 10-phenylanthracene-9-boronic acid, 10-phenylantrhacen-9-yl boronic acid, boronic acid, 10-phenyl-9-anthracenyl, pubchem19639, 9-borono-10-phenylanthracene, 10-phenylanthracene-9-ylboronic acid, 10-phenyl-9-anthryl boronic acid | |
RVPCPPWNSMAZKR-UHFFFAOYSA-N | |
(10-phenylanthracen-9-yl)boronic acid | |
22247164 | |
98% | |
10-Phenylanthracene-9-boronic acid |
C20H15BO2 | |
250 mg | |
Light sensitive | |
Soluble in methanol. | |
OB(O)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12 | |
298.15 | |
298.15 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only