Mopec™ #10 Sterile Scalpel Blades
High-quality, sterile blades used for anatomic pathology or autopsy.
$413.68 - $422.97
Specifications
Product Type Scalpel
Size # 10
Disposable Single-use
Sterility Sterile
Quantity 100 Blades/Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Blade Material Price Quantity  
Catalog Number Mfr. No. Blade Material Price Quantity  
22-445-434
View Documents
Mopec™
AJ203
Stainless Steel
Pack of 100 Each for $422.97
22-445-383
View Documents
Mopec™
AJ170
Carbon Steel
Pack of 100 Each for $413.68
Description
Description
  • Individually wrapped
  • Fits #3, #3L and #5 scalpel handles
Specifications
Specifications
Scalpel
Single-use
100 Blades/Pk.
# 10
Sterile
No
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'